Learn More
Bohlender™ BOLA™ Polypropylene Tube End Fittings
Different colors for better distinction. Polypropylene Tube fittings with polyamide washers offer very good chemical resistance and are available in a variety of sizes.
Brand: Bohlender™ F702-32
Additional Details : Weight : 0.50000kg
Description
- Colored tube end fittings made of PP
- Male thread UNF ¼ in. 28 G
- Washers made of PA are included in delivery
- Supplied in packs of 10
Compliance
FDA
Specifications
Tube End Fitting | |
Polypropylene, PA | |
10/Pk. | |
UNF 1/4 in.; 28 G | |
100°C |
FDA | |
0°C | |
1.6 I.D. x 3.2mm O.D. tubing | |
Green |