missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-[(11bR)-3H-binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethyl)amine, ≥97%, Alfa Aesar™

Brand:  Alfa Aesar H60842.03

Additional Details : CAS Number : 851870-89-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446046

  • 882.00 EUR / 1g


Estimated Shipment date
from Supplier 20-11-2019
Add to basket

Specifications

Chemical Identifiers

851870-89-8
693.811
VMLOJVKKOABMEO-UHFFFAOYSA-N
11216155
C48H41NP2
MFCD09750438
bis 2-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethyl amine, 2,2'-bis r-1,1'-binaphthyl-2,2'-dimethyl phosphino diethylamine, 2,2'-bis s-1,1'-binaphthyl-2,2'-dimethyl phosphino diethylamine, bis 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, bis r-2-4,5-dihydro-3h-binaphtho 1,2-c:2',1'-e-phosphepino ethyl amine, bis s-2-4,5-dihydro-3h-binaphtho 1,2-c:2',1'-e-phosphepino ethyl amine, bis 2-4,5-dihydro-4-phospha-3h-cyclohepta 2,1-a:3,4-a' dinaphthalene-4-yl ethyl amine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethyl 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 23 ,2,4,6,8,10,15,17,19,21-decaen-13-yl ethyl amine
C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCNCCP5CC6=C(C7=CC=CC=C7C=C6)C8=C(C5)C=CC9=CC=CC=C98)C=CC1=CC=CC=C14

Specifications

1g
Air Sensitive
851870-89-8
MFCD09750438
VMLOJVKKOABMEO-UHFFFAOYSA-N
693.811
693.79
≥97%
Bis(2-[(11bR)-3H-binaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethyl)amine
White
C48H41NP2
bis 2-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethyl amine, 2,2'-bis r-1,1'-binaphthyl-2,2'-dimethyl phosphino diethylamine, 2,2'-bis s-1,1'-binaphthyl-2,2'-dimethyl phosphino diethylamine, bis 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethyl amine, bis r-2-4,5-dihydro-3h-binaphtho 1,2-c:2',1'-e-phosphepino ethyl amine, bis s-2-4,5-dihydro-3h-binaphtho 1,2-c:2',1'-e-phosphepino ethyl amine, bis 2-4,5-dihydro-4-phospha-3h-cyclohepta 2,1-a:3,4-a' dinaphthalene-4-yl ethyl amine, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethyl 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 23 ,2,4,6,8,10,15,17,19,21-decaen-13-yl ethyl amine
C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CCNCCP5CC6=C(C7=CC=CC=C7C=C6)C8=C(C5)C=CC9=CC=CC=C98)C=CC1=CC=CC=C14
11216155
Solid
Videos
SDS Documents