missing translation for 'onlineSavingsMsg'
Learn More

BioWhittaker™ Water for Cell Culture, Lonza™

Catalog Number 11650001
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Water for Injection (WFI) system for culture applications. BioWhittaker™ Water for Cell Culture is manufactured aseptically under cGMP conditions using a validated sterile filtration method.

  • Method for the production of WFI includes:
  • Ultra filtration
  • Reverse osmosis
  • Mixed bed deionizers
  • Hot recirculating loop and cooled at point of use
  • System is validated and monitored for bioburden, endotoxin, total organic carbons and conductivity
Quantity 500 mL
Chemical Name or Material Water
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'