Learn More
Agilent BioTek Microplate Washer Flat Magnets
For insertion into washer biomagnetic separation carriers
1565.00 EUR
Specifications
For Use With (Equipment) | 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer |
---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12611795
|
Agilent BioTek
7103016 |
96-Well Flat Magnet |
1565.00 EUR
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In flat-bottom well, beads pulled to band across well bottom. In round-bottom well, beads pulled to button at well bottom.
Order Info
For use with: 405 LS , 405 Touch, EL406, ELx405, ELx50 washers
Specifications
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer |