Learn More
Invitrogen™ Biocytin Alexa Fluor™ 488 (Alexa Fluor™ 488 Biocytin, Disodium Salt)
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
Description
The cell-impermeant, fixable, polar tracer Alexa Fluor™ 488 biocytin combines the green-fluorescent Alexa Fluor™ 488 fluorophore with biotin and an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The biotin allows for detection and subsequent amplification with enzyme-modified streptavidins including horseradish (HRP) and alkaline phosphatase (AP) that can be used with TSA and ELF technologies, respectively.
Specifications
Specifications
| Quantity | 250 μg |
| Content And Storage | Store at room temperature and protect from light. |
| Label or Dye | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Product Line | Alexa Fluor™ |