Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry
Brand: Applied Biosystems™ 4337458
Additional Details : Weight : 0.00450kg
Includes: 10 x 20mL Tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C)
Description
- Improve the quality of your results for a wide range of sequencing applications
- Optimized for long read lengths
- Better dye mobility characteristics
- Improved performance reading through GT rich regions
- Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
- Enhance your productivity and reduce costs
Order Info
Each
Specifications
• 10 × 20-ml tubes of BigDye™ Terminator v3.1 Ready Reaction Mix • 4 tubes M13 (-21) Primer • 4 tubes pGEM Control DNA • 1 × 233-ml tube of 5X Sequencing Buffer (may be stored at 4°C) Store kit at -15°C to -25°C, and avoid repeated freeze-thaw cycles. |
|
12-well Strips, 8-well Strip, 96- or 384-well Plate | |
Standard Sequencing | |
25,000 | |
Sequencing Buffer | |
BAC DNA, Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA |
Cycle Sequencing Kit, 25,000 rxns, Includes 10 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C) | |
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, Veriti Thermal Cycler, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer | |
10 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C) | |
BigDye™ Terminator, BigDye™ | |
Fluorescent Dye Terminator Sequencing |
For Research Use Only. Not for use in diagnostic procedures.