missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Rack Slanter
Tilts test tube rack to 25°, 35° or 45°
Brand: Bel-Art™ F18890-0100
20.94 EUR valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
- Increases the surface area of agar inside test tubes
- Decreases hand strain when working with pipetters by adjusting the rack angle
- Fits racks up to 11.56L x 9.56W x 3.25 in. H (29.3 x 24.2 x 8.3cm)
- Base legs have predetermined slots that stand slides in and out of to easily change angles
- Rigid polypropylene construction
- Steam autoclavable at 121°C (250°F)
- Also for use with the Mini-Magnifier (Mfr. No. 378650000) to aid in the viewing of Petri dishes
Specifications
Tube Rack Slanter | |
Autoclavable | |
Polypropylene |