Learn More
SP Bel-Art™ Bel-Art Products Sterileware Cupped Powder Spatulas
Available in both white and blue, these sterile spatulas are ideal for surface and general sampling of powders
Brand:  SP Bel-Art™ F36932-0150
Additional Details : Weight : 32.00000kg
 View more versions of this product
Product Code. 16896906
  • 220.00 EUR / Pack of 100
Estimated Shipment: 22-07-2024
Description
  • Closed end keeps powders secure within spatula
  • Chose contrasting color for easy visibility if left in material
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • White high impact polystyrene (HIPS), or blue polypropylene (PP) spatulas heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
100 Pk.
Blue
1.8 cm
20 mL
15 cm
23 cm