Learn More
Berkshire™ BCR™ Cleanroom Bond Paper, Wire Bound 36 Hole Punched
Our BCR™ Bond is made with the purest quality cellulose and is an excellent choice for use in copiers and laser printers
Brand: Berkshire™ B85WH36HP25010
284.29 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Additional Details : Weight : 13.50000kg
Description
Specifications
ISO Class 3 - 7, Cleanroom Class 1 - 10,000, EU Grade B/C/D | |
Cleanroom bond paper | |
850 g |
Wire bound, 36 hole punched | |
A4 |