Learn More
Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks
Ensure the purity of distilled water with the copper and tin-crafted reservoirs of Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks.
Brand: Thermo Scientific™ B3043
Additional Details : Weight : 10.00000kg
Description
Ensure the purity of distilled water with the copper and tin-crafted reservoirs of Thermo Scientific™ Barnstead™ Classic Stills Storage Tanks. Select from 10 to 200 gallon reservoir options. All reservoirs include a removable cover, water level sight glass and a tin-coated draw-off faucet with serrated hose nipple. Connections allow for optional accessories. Tanks require a floor stand or wall bracket (10 and 25 gallon reservoirs only) for proper installation.
Ordering Information:
Replacement consumables include UV Lamp (04141), Ventgard Air Filter (H3120), replacement Water Seal (H3130) and Ventgard Filter Element (25001); all sold separately.
Requires:
Floor stand (all sizes) or wall bracket (10- and 25 gal. reservoirs only) are required for secure installation. Ventgard Air Filter/Water Seal (H3111), and UV Light Assembly (H4005) are also required to protect pure water during storage.
Specifications
Storage Tank | |
10 gal Classic Still Reservoir | |
38 L | |
43 cm | |
N/A | |
74 cm | |
26 cm |
Classic Still | |
10 gal. | |
17 in. | |
0.5 in. (distribution) (NPT) | |
29 in. | |
10.25 in. |