Learn More
Bacteriological Peptone, Ultrapure, Thermo Scientific™
High grade peptone prepared specifically for general bacteriological purposes.
Brand: Thermo Scientific J20048.=A
Additional Details : CAS Number : 73049-73-7 Weight : 3.00000kg
Description
- Format: Powder
- CAS: 73049-73-7
- MDL Number: MFCD00131829
Specifications
Powder | |
Bacteriological Peptone | |
Plastic Bottle |
Ultrapure | |
For General Bacteriological Purposes | |
5 lb. |
RUO – Research Use Only