Learn More
Motic™ Epi-Fluorescence Attachment with Filter Cassette, BA410E, BA310E, AE31E
Easily integrated into the BA410 arm. Motic™ Epi-Fluorescence Attachment with Filter Cassette, BA410E, BA310E, AE31E accommodates four filter cassettes for excitation observation.
Brand: Motic™ 1101000202161
Additional Details : Weight : 0.01000kg
Description
- Four cassette capacity.
- Slider format for cassette change.
Specifications
Epi-fluorescence Attachment | |
BA410E, BA310E, AE31E Microscopes | |
Filter cassette |