Learn More
Azlon™ Spouted HDPE Bucket
Spouted HDPE buckets for easy pouring and with moulded graduations
Brand:  Azlon BWX114
40.76 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.94000kg
 View more versions of this product
Product Code. 12064588
  • 42.90 EUR / Each
Estimated Shipment: 12-08-2024
Description
    • Handle grip for easy lifting and carrying
    • Choice of white or black
    • Moulded graduation
Specifications
Specifications
Spouted Bucket
0.5 L
No
330 mm
1 Ea.
17 L
HDPE
White
315 mm