missing translation for 'onlineSavingsMsg'
Learn More

AY 9944 Dihydrochloride, Thermo Scientific Chemicals

Catalog Number 15486709
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
366-93-8
C22H28Cl2N2·2HCl, C22H30Cl4N2
1,4-Bis(2-chlorobenzylaminomethyl)cyclohexane dihydrochloride
This item is not returnable. View return policy
366-93-8
C22H28Cl2N2·2HCl, C22H30Cl4N2
1,4-Bis(2-chlorobenzylaminomethyl)cyclohexane dihydrochloride

Inhibitor of hedgehog (Hh) signaling

Inhibitor of hedgehog (Hh) signaling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 10 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) Inhibitor of hedgehog (Hh) signaling
Formula Weight 464.30g/mol
Sensitivity Hygroscopic
Product Type Signal Transduction Reagent
Form Solid
InChI Key NRVIEWRSGDDWHP-UHFFFAOYSA-N
SMILES [H+].[H+].[Cl-].[Cl-].ClC1=CC=CC=C1CNCC1CCC(CNCC2=CC=CC=C2Cl)CC1
IUPAC Name dihydrogen [(2-chlorophenyl)methyl]({[4-({[(2-chlorophenyl)methyl]amino}methyl)cyclohexyl]methyl})amine dichloride
Molecular Weight (g/mol) 464.30
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'