Learn More
Axygen™ AxyMats™ Sealing Mat for 750uL 96 Round Well Plate and ELISA Plate
Minimize evaporation in wide range of temperatures in PCR and storage applications
Brand: Axygen™ AM-750UL-RD
Additional Details : Weight : 1.09000kg
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Natural | |
Non-sterile | |
96 | |
10/Pk. | |
Round |
Silicone | |
Autoclavable | |
50/Cs. | |
Sealing Mat | |
96-well Round Well Plate and ELISA Plate, 750 μL |