Learn More
Invitrogen™ AURKB (Aurora B) Recombinant Human Protein
The Aurora kinases associate with microtubules during chromosome movement and segregation.
Brand: Invitrogen™ PV6131
Additional Details : Weight : 0.01000kg
Description
AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers.
Specifications
NP_004208 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
AURKB (Aurora B) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
43.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
Aurora Kinase Family | |
Recombinant | |
Baculovirus |