missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AURKA (Aurora A), His Tag Recombinant Protein
Human AURKA (Aurora A), His Tag Recombinant Protein
654.00 EUR - 7890.00 EUR
Specifications
Accession Number | NP_940839 |
---|---|
Gene Alias | AURKA (Aurora A), STK6 |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10379062
|
Invitrogen
PV4067 |
1mg |
7890.00 EUR
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11573467
|
Invitrogen
PV3612 |
10μg |
654.00 EUR
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AURKA (Aurora A) is a serine⁄threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer.Specifications
NP_940839 | |
Human | |
Baculovirus |
AURKA (Aurora A), STK6 | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.