Learn More
Atrial Natriuretic Peptide/ANP Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand:  R&D Systems AF3366-SP
Additional Details : Weight : 0.00970kg
Product Code. 15701994
  • 256.50 EUR / 25µg
Estimated Shipment: 24-06-2024
to see stock.
Description
Description
Atrial Natriuretic Peptide/ANP Polyclonal specifically detects Atrial Natriuretic Peptide/ANP in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.