missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATF1 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1475.00 EUR - 2430.00 EUR
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | activating transcription factor 1 |
Gene Alias | EWS-ATF1,FUS/ATF-1,TREB36 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023485
|
Abnova™
FS0048.100UL |
100 μL |
1475.00 EUR
100µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033485
|
Abnova™
FS0048.200UL |
200 μL |
2430.00 EUR
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
EWS-ATF1,FUS/ATF-1,TREB36 | |
466 | |
Liquid |
Tube | |
activating transcription factor 1 | |
ATF1 | |
DNA/RNA |