missing translation for 'onlineSavingsMsg'
Learn More

Ascorbic Acid USP, Macron Fine Chemicals™

Product Code. p-3176980
Click to view available options
Quantity:
500 g
Packaging:
Glass
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15507934

Brand: Avantor Macron 185210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Packaging Glass
Quantity 500 g
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Formula Weight 176.13
Grade USP
Chemical Name or Material Ascorbic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.