Learn More
Thermo Scientific™ ART™ Non-Filtered Gel 200 Universal-Fit Pipette Tips
For loading specific types of gels
Brand: Thermo Scientific™ 3690
Additional Details : Weight : 0.36000kg
Description
For loading specific types of gels
- Narrow, extended tip fits easily between glass plates and into wells of electrophoresis gels
- Round end
- Universal Tip, fits most brands of pipettes
- ART™ non-filtered tips deliver the same proven quality, performance and reliability as ART™ barrier
- Non-filtered tip
- ART™ tips are certified to be free of DNA, RNase, DNase, ATP, Endotoxins, and Pyrogens
- Lift-off lid racks provide access to all tips, great for multichannel pipetting
Specifications
ART 200 Gel, Non-filtered, Non-sterile, Bulk | |
Non-Filtered | |
Clear | |
ART 200 Gel | |
loading electrophoresis gels -PAGE and agarose | |
Bulk | |
bulk | |
Single-use | |
ART |
200 μL | |
Non-sterile | |
Autoclavable | |
DNase, RNase, DNA, ATP, PCR Inhibitor, Endotoxin, and Pyrogen Free | |
gel loading tip | |
Gel Loading | |
10000 | |
Universal Fit | |
Virgin Polypropylene |