missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Mouse anti-Canine, Human, Clone: 1H6C3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66739-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ADP-ribosylation factor-like family. The encoded protein is a small GTPase that contains both N-terminal and C-terminal guanine nucleotide-binding motifs. This protein is localized in the cilia and plays a role in cilia formation and in maintece of cilia. Mutations in this gene are the cause of Joubert syndrome 8. Alternate splicing results in multiple transcript variants.Specifications
ARL13B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3SXY8 | |
ARL13B | |
ARL13B Fusion Protein Ag12031 | |
20 μL | |
Primary | |
Human, Canine | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1H6C3 | |
Unconjugated | |
ARL13B | |
ARL13B, ARL2 like protein 1, ARL2L1, DKFZp686E2075, DKFZp686L2472, DKFZp686M2074, DKFZp761H079, JBTS8 | |
Mouse | |
Protein A | |
RUO | |
200894, 487934 | |
-20°C | |
Liquid |