Learn More
Hirschmann™ Class AS Graduated Pipettes with Amber Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes with Amber Graduation come with a cotton plug end as standard.
Brand:  Hirschmann™ 1100235
Additional Details : Weight : 0.29610kg
 View more versions of this product
Product Code. 11776632
  • 82.20 EUR / Pack of 12
Estimated Shipment: 04-06-2024
Description
  • Main point ring graduation
  • Graduated not to tip, zero on top
  • Acc. to DIN EN ISO 835
  • Conformity certified, with dated batch identification
Specifications
Specifications
AS
Yes
0.1 mL
12 Pack
1 mL
Graduated Pipet
AR-GLAS
Amber
Durable amber stain diffusion
0.007 mL
360 ±5 mm