Learn More
Lovibond™ Aqua Comparitor Color Discs: Nitrate
For testing water with the Aqua Comparator
26.10 EUR
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
Type Nitrate Color Disc
View More Specs
Products 1
Product Code Brand Type Range Price Quantity & Availability  
Product Code Brand Type Range Price Quantity & Availability  
11745548
View Documents Promotion Details
Lovibond™
146300
Nitrate Color Disc 0 to 0.5mg/L
26.10 EUR
Each
View alternative products
Save Up to 
»
Estimated Shipment: 10-06-2024
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for nitrates using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
Testing water with the Aqua comparator™
Aqua comparator™
Nitrate Color Disc