Learn More
Lovibond™ Aqua Comparitor Color Discs: Nitrate
For testing water with the Aqua Comparator
26.10 EUR
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Type | Nitrate Color Disc |
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for nitrates using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
Nitrate Color Disc |