Learn More
USP47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530661
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, mouse brain. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (85%), Xenopus laevis (86%), Chicken (94%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
USP47 Polyclonal antibody specifically detects USP47 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
USP47 | |
Polyclonal | |
Unconjugated | |
USP47 | |
4930502N04Rik; A630020C16Rik; deubiquitinating enzyme 47; Trf (TATA binding protein-related factor)-proximal homolog; TRFP; Ubiquitin carboxyl-terminal hydrolase 47; ubiquitin specific peptidase 47; ubiquitin specific protease 47; ubiquitin thioesterase 47; ubiquitin thiolesterase 47; ubiquitin-specific-processing protease 47; Usp47 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55031, 74996 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8BY87, Q96K76 | |
USP47 | |
Recombinant fragment corresponding to a region within amino acids 1186 and 1375 of Human USP47 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |