Learn More
SFTPB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542000
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PGALQARPGP HTQDLSEQQF PIPLPYCWLC RALIKRIQAM IPKGALAVAV Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 92%; Rabbit: 85%; Rat: 100%; Sheep: 79%
SFTPB Polyclonal antibody specifically detects SFTPB in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SFTPB | |
Polyclonal | |
Unconjugated | |
SFTPB | |
18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
192155, 20388, 6439 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P07988, P22355, P50405 | |
SFTPB | |
Synthetic peptide directed towards the middle region of human SFTPB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |