Learn More
SREBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1338
Additional Details : Weight : 0.01000kg
Description
PA1-338 detects SREBP 2 in human, mouse and rat samples. The PA1-338 immunogen is a synthetic peptide corresponding to residues A(843) I S W L Q G D D A A V R S H(857) of rat SREBP 2. This sequence is identical in human and differs by 1 a.a. in mouse. PA1-338 immunizing peptide (Cat. # PEP-275) is available for use in neutralization and control experiments.
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.Specifications
SREBP2 | |
Polyclonal | |
Unconjugated | |
SREBF2 | |
AI608257; bHLHd2; Class D basic helix-loop-helix protein 2; EMBL:AAI01903.1}; lop13; nuc; Processed sterol regulatory element-binding protein 2; srebf2; srebf2 {ECO:0000312; Srebf2_retired; SREBP2; SREBP-2; SREBP2gc; sterol regulatory element binding factor 2; sterol regulatory element binding protein 2; sterol regulatory element binding transcription factor 2; sterol regulatory element-binding protein 2; Sterol regulatory element-binding transcription factor 2; zgc:158371 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20788, 300095, 6721 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q12772, Q3T1I5, Q3U1N2 | |
SREBF2 | |
Synthetic peptide corresponding to residues A(843) I S W L Q G D D A A V R S H(857) of rat SREBP 2. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |