missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA19-9 Monoclonal Antibody (121SLE), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119211
Additional Details : Weight : 1.24740kg
Description
For IHC (P), heat mediated antigen retrieval in sodium citrate is recommended.
CA19-9 Monoclonal antibody specifically detects CA19-9 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
CA19-9 | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
Mouse | |
sequential chromatography | |
RUO | |
Human | |
Antibody | |
IgM |
ELISA, Immunohistochemistry (Paraffin) | |
121SLE | |
Unconjugated | |
CA19 9; CA19.9; CA-19.9 | |
Immunoprecipitate obtained after immunodiffusion of MAb 19-9 and mucins isolated from an ovarian cyst of a 0Le(a+b-) patient. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |