missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2CA/PP2CB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112395
Additional Details : Weight : 0.01000kg
Description
PP2CA/PP2CB Polyclonal antibody specifically detects PP2CA/PP2CB in Bovine, Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PP2CA/PP2CB | |
Polyclonal | |
PBS with 0.08% sodium azide | |
O62829, O62830, O75688, P20650, P35813, P35815, P36993, P49443 | |
PPM1A, Ppm1b | |
Synthetic peptide. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Ppm1b | |
2310003C21Rik; 2900017D14Rik; AI427932; AU017636; FLJ42306; IA; MGC9201; MMPa-2; Mpp alpha; MPPa-1; Pp2c1; Pp2c2; PP2CA; PP2Calpha; PP2C-ALPHA; PP2CB; PP2CBETA; PP2C-beta; PP2C-beta-X; PPC2BETAX; Ppm1a; PPM1B; PPM1B protein; Pppm1a; Pppm1b; Protein phosphatase 1A; protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform; protein phosphatase 1A, magnesium dependent, alpha isoform; protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; protein phosphatase 1B, magnesium dependent, beta isoform; protein phosphatase 2C alpha; protein Phosphatase 2C beta; protein phosphatase 2C isoform alpha; Protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; protein phosphatase IA; Protein phosphatase type 1A (formely 2C) Mg-dependent alpha isoform; Protein phosphatase type 1A (formely 2C), Mg-dependent, alpha isoform; Protein phosphatase type 1B (formely 2C) Mg-dependent beta isoform; Protein phosphatase type 1B (formely 2C), | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
19042, 19043, 24666, 24667, 281994, 281995, 5494, 5495 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |