Learn More
PLA2G3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532987
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%); Marmoset, Mouse, Rat, Dog, Hamster (94%); Elephant, Panda, Rabbit (88%); Bovine, Horse, Pig (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
PLA2G3 Polyclonal antibody specifically detects PLA2G3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
PLA2G3 | |
Polyclonal | |
Unconjugated | |
PLA2G3 | |
giii; GIII sPLA2; GIII-SPLA2; Group 3 secretory phospholipase A2; group III secreted phospholipase A2; Group III secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 3; phosphatidylcholine 2-acylhydrolase GIII; phospholipase A2 group III; phospholipase A2, group III; PLA2G3; SPLA2III; sPLA2-III | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50487 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NZ20 | |
PLA2G3 | |
Synthetic 16 amino acid peptide from internal region of human PLA2G3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |