Learn More
Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MN1020B
Additional Details : Weight : 2.72160kg
Description
MN1020B targets PHF-tau (Ser202/Thr205) in IF, ELISA, and IHC (P) applications and shows reactivity with Human samples. The MN1020B immunogen is partially purified human PHF-Tau. MN1020B detects PHF-tau (Ser202/Thr205)a which has a predicted molecular weight of approximately 79 kDa.
Phospho-Tau (Ser202, Thr205) Monoclonal antibody specifically detects Phospho-Tau (Ser202, Thr205) in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
Phospho-Tau (Ser202, Thr205) | |
Monoclonal | |
0.1 mg/mL | |
PBS with no preservative | |
P10636 | |
MAPT | |
Partially purified human PHF-Tau | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
AT8 | |
Biotin | |
MAPT | |
AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287) | |
Mouse | |
Protein A | |
RUO | |
4137 | |
4° C | |
Liquid |