missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI1 Monoclonal Antibody (3F6H5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515776
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human ovary tissue lysate.
ORAI1 Monoclonal antibody specifically detects ORAI1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ORAI1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5M848, Q8BWG9, Q96D31 | |
Orai1 | |
A 16 amino acid peptide from near the carboxy terminus of human ORAI1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
3F6H5 | |
Unconjugated | |
Orai1 | |
calcium release-activated calcium channel 1; calcium release-activated calcium channel protein 1; calcium release-activated calcium modulator 1; CRACM1; D730049H07Rik; FLJ14466; hypothetical protein FLJ14466; hypothetical protein LOC402892; im:7146264; IMD9; ORAI calcium release-activated calcium modulator 1; ORAI calcium release-activated calcium modulator 1b; orai1; orai-1; orai1b; ORAT1; protein orai-1; RGD1311873; TAM2; TMEM142A; Transmembrane protein 142A; zFLJ14466; zgc:109721 | |
Mouse | |
Protein A | |
RUO | |
109305, 304496, 84876 | |
-20°C | |
Liquid |