missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOC4L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541906
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: CRVLVHRPHG PELDADPYDP GEEDPAQSRA LESSLWELQA LQRHYHPEVS Sequence homology: Cow: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 86%
NOC4L Polyclonal antibody specifically detects NOC4L in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NOC4L | |
Polyclonal | |
Unconjugated | |
NOC4L | |
AI326906; NET49; NOC4; NOC4 protein homolog; NOC4L; NOC4-like protein; nucleolar complex associated 4 homolog; nucleolar complex associated 4 homolog (S. cerevisiae); nucleolar complex protein 4 homolog; Nucleolar complex-associated protein 4-like protein; RGD1310661; UTP19 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79050 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BVI4 | |
NOC4L | |
Synthetic peptide directed towards the C-terminal of human NOC4L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |