Learn More
MYL7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530789
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, HeLa, mouse heart, rat heart. Predicted reactivity: Mouse (95%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MYL7 Polyclonal antibody specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MYL7 | |
Polyclonal | |
Unconjugated | |
Myl7 | |
MLC2a; MLC-2a; MLC-2alpha; MYL2A; MYL7; Mylc2a; myosin light chain 2a; myosin light chain 7; myosin light chain, regulatory A; myosin regulatory light chain 2, atrial isoform; Myosin regulatory light chain 7; myosin, light chain 7, regulatory; myosin, light polypeptide 7, regulatory; RLC-A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17898, 289759, 58498 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
3.11 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q01449, Q9QVP4 | |
Myl7 | |
Recombinant fragment corresponding to a region within amino acids 1 and 175 of Human MYL7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |