missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 594, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A21135
241.50 EUR valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
Product will be shipped at Room Temperature.
Mouse IgG2a Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG2a Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Mouse IgG2a Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 594 | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Mouse IgG2a | |
500 μg | |
Secondary | |
4° C, store in dark | |
IgG |