missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-G Monoclonal Antibody (MEM-G/1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119219
Additional Details : Weight : 0.10000kg
Description
This antibody reacts with denaturated HLA-G heavy chain. Immunohistochemistry (Paraffin): Incubation: 1 h at RT.
HLA-G Monoclonal antibody specifically detects HLA-G in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
HLA-G | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P17693 | |
HLA-G | |
Recombinant human HLA-G denaturated heavy chain. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
MEM-G/1 | |
Unconjugated | |
HLA-G | |
acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 | |
Mouse | |
Protein A | |
RUO | |
3135 | |
4° C, do not freeze | |
Liquid |