missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 15 Monoclonal Antibody (LHK15), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA190929
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Skin.
Cytokeratin 15 Monoclonal antibody specifically detects Cytokeratin 15 in Bovine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Cytokeratin 15 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P19012, Q61414, Q6IFV3 | |
KRT15 | |
A 17-mer synthetic peptide from the C-terminus of Keratin 15 | |
500 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
LHK15 | |
Unconjugated | |
KRT15 | |
AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012} | |
Mouse | |
Protein A | |
RUO | |
16665, 287700, 3866, 507464 | |
4° C, do not freeze | |
Liquid |