missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A Monoclonal Antibody (F.674.9), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514891
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
HNF4A Monoclonal antibody specifically detects HNF4A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
HNF4A | |
Monoclonal | |
52 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P41235 | |
HNF4A | |
Synthetic peptide corresponding to the sequence of human HNF4a | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
F.674.9 | |
Unconjugated | |
HNF4A | |
alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3172 | |
-20°C | |
Liquid |