Learn More
FMR1, Mouse, Clone: 2D4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FMR1.
Brand: Abnova H00002332-M01.100ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene binds RNA and is associated with polysomes. The encoded protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat (CGG) in the 5' UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure (POF1). [provided by RefSeq]
Sequence: ATKDTFHKIKLDVPEDLRQMCAKEAAHKDFKKAVGAFSVTYDPENYQLVILSINEVTSKRAHMLIDMHFRSLRTKLSLIMRNEEASKQLESSRQLASRFHSpecifications
FMR1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002024 | |
FMR1 | |
FMR1 (NP_002015, 121 a.a. ∼ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, KnockDown, Western Blot | |
2D4 | |
Mouse monoclonal antibody raised against a partial recombinant FMR1. | |
FMR1 | |
FMRP/FRAXA/MGC87458/POF/POF1 | |
Mouse | |
Affinity chromatography | |
RUO | |
2332 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |