Learn More
Fibrillarin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529801
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, H1299, Molt-4, Neuro2A, PC-12, A549, A549 nuclear extract, Jurkat, Jurkat nuclear extract, SK-N-SH, SK-N-SH nuclear extract, IMR32, IMR32 nuclear extract. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (89%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Fibrillarin Polyclonal antibody specifically detects Fibrillarin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Fibrillarin | |
Polyclonal | |
Unconjugated | |
fbl | |
34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14113, 2091, 292747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P22087, P22509, P35550 | |
fbl | |
Recombinant fragment corresponding to a region within amino acids 75 and 321 of Human Fibrillarin | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |