Learn More
Ferritin Monoclonal Antibody (523), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514735
Additional Details : Weight : 0.01000kg
Description
MA5-14735 detects Ferritin in ELISA, IP, IF, and RIA applications and shows reactivity to human samples. The MA5-14735 Immunogen is human liver ferritin. MA5-14735 detects ferritin which has a predicted molecular weight of approximately 19kDa. Product MA514735 is a smaller package size of MIF2503 (formerly sold as a Seradyn product).
Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA).Specifications
Ferritin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02792, P02794 | |
FTH1, FTL | |
Human liver ferritin | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
523 | |
Unconjugated | |
FTH1 | |
apoferritin; cell proliferation-inducing gene 15 protein; Ferritin H subunit; ferritin heavy chain; ferritin heavy chain 1; Ferritin heavy chain, N-terminally processed; Ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3; Ferritin subunit H; ferritin, heavy polypeptide 1; ferritin, light polypeptide; FHC; frih; fril; FTH; Fth1; FTHL6; FTL; HFE5; H-ferritin; HFt; L apoferritin; LFTD; MFH; MGC71996; NBIA3; OK/SW-cl.84; PIG15; placenta immunoregulatory factor; PLIF; proliferation-inducing protein 15 | |
Mouse | |
Protein A | |
RUO | |
2495, 2512 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |