Learn More
Dynamin Mouse, Unlabeled, Clone: 41, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610245
Additional Details : Weight : 0.01000kg
Description
Dynamin is a nerve terminal phosphoprotein with intrinsic GTPase activity which plays an important role in endocytosis. This GTPase activity is stimulated in vitro by microtubules, SH3 domain-containing proteins, phospholipids, and Protein Kinase C (PKC)-mediated phosphorylation. There are at least two distinct dynamin genes in mammals, which show 79% identity, encoding proteins Dynamin I and Dynamin II. Dynamin I is expressed almost exclusively in the central nervous system while Dynamin II expression is ubiquitous. Depolarization stimulates synaptic vesicle recycling and Dynamin I is subsequently dephosphorylated. This rapid dephosphorylation is mediated by calcineurin which acts as a switch for depolarization-initiated synaptic vesicle endocytosis. The C-terminus of dynamin contains two clusters of proline-rich SH3 domain binding proline motifs which interact with known SH3 domain proteins during tyrosine kinase receptor activation. It is reported that clone 41 detects both dynamin I and dynamin II.
Specifications
Dynamin | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Canine, Chicken, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
41 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.