missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OxPhos Complex IV subunit IV Monoclonal Antibody (20E8C12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A21348
Additional Details : Weight : 0.00450kg
Description
OxPhos Complex IV subunit IV Monoclonal antibody specifically detects OxPhos Complex IV subunit IV in Bovine, Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot.Specifications
OxPhos Complex IV subunit IV | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
Mouse | |
purified | |
RUO | |
Bovine, Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG2a κ |
Western Blot | |
20E8C12 | |
Unconjugated | |
COX4; COX4-1; COXIV; COX IV-1; cytochrome c oxidase polypeptide IV; cytochrome c oxidase subunit 4 isoform 1 mitochondrial | |
Bovine Complex IV (native holoenzyme protein, purified from liver). | |
100 μL | |
Primary | |
4° C | |
Liquid |