Learn More
CD97 Monoclonal Antibody (MEM-180), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516825
354.68 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4oC in the dark. Once reconstituted, this product has a shelf life of approx. 18 months. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD97 Monoclonal antibody specifically detects CD97 in Human samples. It is validated for Flow Cytometry.Specifications
CD97 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
P48960 | |
ADGRE5, CD97 | |
CD97 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
MEM-180 | |
APC | |
ADGRE5 | |
AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1 | |
Mouse | |
Protein A | |
RUO | |
976 | |
4° C, store in dark, DO NOT FREEZE! | |
Lyophilized |