Learn More
CD93 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA546921
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse C1q R1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD93 Polyclonal antibody specifically detects CD93 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
CD93 | |
Polyclonal | |
Unconjugated | |
Cd93 | |
6030404G09Rik; AA145088; Aa4; AA4.1; AW555904; C1q receptor; C1q receptor 1; C1q/MBL/SPA receptor; C1qR; C1qR(P); C1qr1; C1qRP; Cd93; CD93 antigen; CD93 molecule; CDw93; Cell surface antigen AA4; complement component 1 q subcomponent receptor 1; complement component 1, q subcomponent, receptor 1; complement component C1q receptor; dJ737E23.1; ECSM3; Ly68; ly-68; Lymphocyte antigen 68; matrix-remodeling-associated protein 4; matrix-remodelling associated 4; MXRA4 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
22918 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NPY3 | |
Cd93 | |
Mouse myeloma cell line NS0-derived recombinant human C1q R1/CD93 Ala24-Lys580 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |