missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545179
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GTYRCTLQDP DGQRNLSGKV ILRVTGCPAQ RKEETFKKYR AEIVLLLALV Sequence homology: Human: 100%; Pig: 91%
CD83 Polyclonal antibody specifically detects CD83 in Human samples. It is validated for Western Blot.Specifications
CD83 | |
Polyclonal | |
Unconjugated | |
CD83 | |
B-cell activation protein; BL11; CD antigen CD83; CD83; CD83 antigen; CD83 antigen (activated B lymphocytes, immunoglobulin superfamily); CD83 molecule; cell surface protein HB15; cell-surface glycoprotein; HB15; hCD83; mCD83 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9308 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q01151 | |
CD83 | |
Synthetic peptide directed towards the C-terminal of human CD83 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |