missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD80, Clone: 4F7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000941-M32.100ug
Additional Details : Weight : 0.00010kg
Specifications
CD80 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005191.2 | |
CD80 | |
CD80 (NP_005182.1, 35 a.a. ∼ 242 a.a) partial recombinant protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
4F7 | |
Mouse monoclonal antibody raised against a partial recombinant CD80. | |
CD80 | |
CD28LG/CD28LG1/LAB7 | |
Mouse | |
Affinity chromatography | |
RUO | |
941 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
For Research Use Only