Learn More
CD70 Monoclonal Antibody (BU69), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517728
Additional Details : Weight : 0.01000kg
Description
MA5-17728 targets CD70/Ki-24 antigen in FACS and IHC (F) applications and shows reactivity with Human samples. The MA5-17728 immunogen is mouse monoclonal antibodies raised against human CD70.
CD70 Monoclonal antibody specifically detects CD70 in Human samples. It is validated for Flow Cytometry.Specifications
CD70 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P32970 | |
Cd70 | |
Waldenstroms macroglobulineamia cell line WM-1 | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
BU69 | |
PE | |
Cd70 | |
CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7 | |
Mouse | |
Protein G | |
RUO | |
970 | |
4° C, do not freeze | |
Liquid |