missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Monoclonal Antibody (FN50), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518150
Additional Details : Weight : 0.01000kg
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry.Specifications
CD69 | |
Monoclonal | |
Alexa Fluor 647 | |
CD69 | |
5830438K24Rik; Activation inducer molecule; activation inducer molecule (AIM/CD69); AI452015; AIM; BL-AC/P26; CD69; CD69 antigen; CD69 antigen (p60, early T-cell activation antigen); CD69 molecule; CLEC2C; C-type lectin domain family 2 member C; C-type lectin domain family 2, member C; EA1; early activation antigen CD69; early lymphocyte activation antigen; early T-cell activation antigen p60; GP32/28; leukocyte surface antigen Leu-23; MLR-3; VEA; Very Early Activation Antigen | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
969 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
FN50 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q07108 | |
CD69 | |
anti-u-stimulated human B lymphocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |