missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-selectin Monoclonal Antibody (1.2B6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA122165
Additional Details : Weight : 1.24740kg
Description
Not suitable for immunohistochemistry (paraffin). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E-selectin Monoclonal antibody specifically detects E-selectin in Human, Porcine samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
E-selectin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P16581, P98110 | |
SELE | |
Human E-Selectin (ELAM-1) | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
1.2B6 | |
Unconjugated | |
SELE | |
CD62 antigen-like family member E; CD62E; e selectin; Elam; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule; endothelial leukocyte adhesion molecule 1; endothelial leukocyte adhesion molecule I; ESEL; E-selectin; LECAM2; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin, endothelial cell | |
Mouse | |
Protein A | |
RUO | |
397508, 6401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |